Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PD-L1 Rabbit anti-Human, liquid, TrueRAB™

Rabbit Recombinant Monoclonal Antibody
Supplier: OriGene TA591005S

Description
PD-L1 (Programmed death receptor ligand 1, B7-H1, CD274) is a 40 kDa transmembrane protein and B7 family member. PD-L1 has been shown to negatively regulate T-cell receptor (TCR) signaling. Upon ligating its receptor, PD-L1 has been reported to decrease TCR-mediated proliferation and cytokine production, and PD-L1 expression was found to be abundant on many murine and human cancers and could be further up-regulated upon IFN-gamma stimulation. Thus, PD-L1 might play an important role in tumor immune evasion. Together with B7.1, PD-L1 can inactivate T cells and this relationship is a focal point of cancer therapeutic treatment. At least two isoforms of PDL-1 are known to exist.Specifications
PD-L1 | |
Recombinant Monoclonal | |
Unconjugated | |
CD274 | |
B7-H, B7H1, PD-L1, PDCD1L1, PDCD1LG1, PDL1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
29126 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
OR-5E4 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9NZQ7 | |
CD274 | |
Synthetic peptide within Human PD-L1 180-230. The exact sequence is proprietary. | |
30 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction