Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD-L1 Rabbit anti-Human, liquid, TrueRAB™
SDP

Catalog No. 501677099
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-7099 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-7099 Supplier OriGene Supplier No. TA591005
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

PD-L1 (Programmed death receptor ligand 1, B7-H1, CD274) is a 40 kDa transmembrane protein and B7 family member. PD-L1 has been shown to negatively regulate T-cell receptor (TCR) signaling. Upon ligating its receptor, PD-L1 has been reported to decrease TCR-mediated proliferation and cytokine production, and PD-L1 expression was found to be abundant on many murine and human cancers and could be further up-regulated upon IFN-gamma stimulation. Thus, PD-L1 might play an important role in tumor immune evasion. Together with B7.1, PD-L1 can inactivate T cells and this relationship is a focal point of cancer therapeutic treatment. At least two isoforms of PDL-1 are known to exist.

Specifications

Antigen PD-L1
Applications Western Blot
Classification Recombinant Monoclonal
Clone OR-5E4
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CD274
Gene Accession No. Q9NZQ7
Gene Alias B7-H, B7H1, PD-L1, PDCD1L1, PDCD1LG1, PDL1
Gene Symbols CD274
Host Species Rabbit
Immunogen Synthetic peptide within Human PD-L1 180-230. The exact sequence is proprietary.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29126
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.