Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PD-L1 Rabbit anti-Human, Mouse, Rat, Clone: 1T10L4, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $492.50
Specifications
Antigen | PD-L1 |
---|---|
Clone | 1T10L4 |
Dilution | Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin, Knockout Validated |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown |
Classification | Monoclonal |
Description
PD-L1 Monoclonal antibody specifically detects PD-L1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDownSpecifications
PD-L1 | |
Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin, Knockout Validated | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
Avelumab, B7-H, B7-H1, B7H1PDCD1L1, CD274 antigenMGC142294, CD274 molecule, PDCD1LG1MGC142296, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-290 of human PD-L1 (Q9NZQ7). MRIFAVFIFMTYWHLLNAFTVTVPKDLYVVEYGSNMTIECKFPVEKQLDLAALIVYWEMEDKNIIQFVHGEEDLKVQHSSYRQRARLLKDQLSLGNAALQITDVKLQDAGVYRCMISYGGADYKRITVKVNAPYNKINQRILVVDPVTSEHELTCQAEGYPKAEVIWTSSDHQVLSGKTTTTNSKREEKLFNVTSTLRINTTTNEIFYCTFRRLDPEENHTAELVIPELPLAHPPNERTHLVILGAILLCLGVALTFIFRLRKGRMMDVKKCGIQDTNSKKQSDTHLEET | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
1T10L4 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown | |
Unconjugated | |
Rabbit | |
Autoimmune Diseases, Cancer, Cellular Markers, Checkpoint signaling, Immune System Diseases, Immunology, Signal Transduction, Tumor Biomarkers, Tumor Suppressors | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
29126 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title