Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD-L1 Rat anti-Mouse, Clone: 10F.9G2, MilliporeSigma™

Catalog No. MABC991MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC991MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC991MI Supplier MilliporeSigma Supplier No. MABC991
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Detect PD-L1 using this rat monoclonal Anti-PD-L1 Antibody, clone 10F.9G2 Function Induction, Immunohistochemistry, and Inhibition studies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PD-L1
Applications Flow Cytometry, Inhibition Assays, Immunohistochemistry
Classification Monoclonal
Clone 10F.9G2
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols CD274,B7h1,Pdcd1l1,Pdcd1lg1,Pdl1
Host Species Rat
Immunogen CHO-mPD-L1 transfectants.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis and Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_068693
Test Specificity Clone 10F.9G2 specifically detects PD-L1 in murine EL4 lymphoma cells and targets the extracellular domain.
Target Species Mouse
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.