Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PD-L2/B7-DC/PDCD1LG2 Antibody (168633) [DyLight 405], Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB1022E
Description
PD-L2/B7-DC/PDCD1LG2 Monoclonal antibody specifically detects PD-L2/B7-DC/PDCD1LG2 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
PD-L2/B7-DC/PDCD1LG2 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
168633 | |
Western Blot, Flow Cytometry, CyTOF-ready | |
B7-DC, bA574F11.2, Btdc, Butyrophilin B7-DC, CD273 antigen, CD273PD-1 ligand 2, MGC142240, PD-1-ligand 2, PDCD1L2MGC142238, PDL2B7DC, PD-L2PDCD1 ligand 2, programmed cell death 1 ligand 2, Programmed death ligand 2 | |
Mouse myeloma cell line NS0-derived recombinant mouse PD-L2/B7-DC, Met1-Arg219, Accession # Q9WUL5 | |
0.1 mL | |
Apoptosis | |
80380 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction