Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PD-L2/B7-DC/PDCD1LG2 Antibody (208), DyLight 594

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290723DL594
Description
PD-L2/B7-DC/PDCD1LG2 Monoclonal specifically detects PD-L2/B7-DC/PDCD1LG2 in Mouse samples. It is validated for ELISA.Specifications
PD-L2/B7-DC/PDCD1LG2 | |
Monoclonal | |
DyLight 594 | |
B7-DC, bA574F11.2, Btdc, Butyrophilin B7-DC, CD273 antigen, CD273PD-1 ligand 2, MGC142240, PD-1-ligand 2, PDCD1L2MGC142238, PDL2B7DC, PD-L2PDCD1 ligand 2, programmed cell death 1 ligand 2, Programmed death ligand 2 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse PD-L2/B7-DC/PDCD1LG2 (Q9WUL5; Met1-Arg219). | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
ELISA | |
208 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
80380 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction