Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PD-L2/B7-DC/PDCD1LG2 Antibody (OTI7B10), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273306B
Description
PD-L2/B7-DC/PDCD1LG2 Monoclonal specifically detects PD-L2/B7-DC/PDCD1LG2 in Human samples. It is validated for Western Blot.Specifications
PD-L2/B7-DC/PDCD1LG2 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
PDCD1LG2 | |
Full length recombinant protein of human PD-L2/B7-DC/PDCD1LG2 (NP_079515) produced in HEK293T cells. | |
0.1 mL | |
80380.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI7B10 | |
Western Blot | |
B7-DC, bA574F11.2, Btdc, Butyrophilin B7-DC, CD273 antigen, CD273PD-1 ligand 2, MGC142240, PD-1-ligand 2, PDCD1L2MGC142238, PDL2B7DC, PD-L2PDCD1 ligand 2, programmed cell death 1 ligand 2, Programmed death ligand 2 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction