Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PD-L2/B7-DC/PDCD1LG2 Antibody (TY25), Allophycocyanin, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB10065332APC
Description
PD-L2/B7-DC/PDCD1LG2 Monoclonal specifically detects PD-L2/B7-DC/PDCD1LG2 in Mouse samples. It is validated for Flow Cytometry.Specifications
PD-L2/B7-DC/PDCD1LG2 | |
Monoclonal | |
APC | |
PBS with 0.05% Sodium Azide | |
PDCD1LG2 | |
B7-DC transfected RAW264.7 cells. | |
0.125 mL | |
Primary | |
NB100-65332 recognizes mouse CD273, a 42kD member of the B7 family, otherwise known as B7-dendritic cell (B7-DC), or programmed death ligand-2 (PD-L2). The expression of CD273 is restricted to a subpopulation of dendritic cells (DCs), and DCs and macrophages that have been stimulated with interferon gamma, GM-CSF or IL-4. CD273 has been identified as a ligand for PD-1(CD279), along with another B7 family member B7-H1 (CD274). Studies suggest that both CD273 and B7-H1 co-stimulate or inhibit T cell proliferation and cytokine production. Antibodies produced by clone TY25 are reported to block the binding of CD273 (B7-DC) to it's receptor. | |
Store at 4C in the dark. | |
IgG2a |
Flow Cytometry | |
TY25 | |
Flow Cytometry | |
B7-DC, bA574F11.2, Btdc, Butyrophilin B7-DC, CD273 antigen, CD273PD-1 ligand 2, MGC142240, PD-1-ligand 2, PDCD1L2MGC142238, PDL2B7DC, PD-L2PDCD1 ligand 2, programmed cell death 1 ligand 2, Programmed death ligand 2 | |
Rat | |
Protein G purified | |
RUO | |
80380 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction