Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PD-L2 (CD273) Mouse anti-Human, Alexa Fluor™ 647, Clone: IPI683.rMab, BD Phrmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 569412
Description
PD-L2 (CD273) Monoclonal antibody specifically detects PD-L2 (CD273) in Human samples. It is validated for Flow cytometry
Specifications
PD-L2 (CD273) | |
Monoclonal | |
Alexa Fluor 647 | |
CD273; PD-L2; PDL2; PDCD1LG2; B7-DC; B7DC | |
Human PD-L2 cDNA | |
25 Tests | |
Primary | |
Human | |
Liquid |
Flow Cytometry | |
IPI683.rMab | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
80380 | |
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 λ1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction