Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDAP1 Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP297989AF647

Catalog No. NB050547


Only null left
Add to Cart

Description

Description

PDAP1 Polyclonal specifically detects PDAP1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

PDAP1
Polyclonal
Immunocytochemistry/Immunofluorescence
HASPP28, PAP1PDGFA-associated protein 1, PAP28 kDa heat- and acid-stable phosphoprotein, PDGFA associated protein 1, PDGF-associated protein
Produced in rabbits immunized with E. coli-derived Human PDAP1 fragment.
0.1 mL
Signal Transduction
11333
Store at 4°C in the dark.
IgG
Immunofluorescence
Alexa Fluor 647
50mM Sodium Borate
Rabbit
Antigen Affinity-purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.