Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDAP1 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297989APC
Description
PDAP1 Polyclonal antibody specifically detects PDAP1 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
PDAP1 | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence | |
HASPP28, PAP1PDGFA-associated protein 1, PAP28 kDa heat- and acid-stable phosphoprotein, PDGFA associated protein 1, PDGF-associated protein | |
Produced in rabbits immunized with E. coli-derived Human PDAP1 fragment. | |
0.1 mL | |
Signal Transduction | |
11333 | |
Store at 4C in the dark. | |
IgG |
Immunocytochemistry | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction