Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDAP1 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP320223MFV610
Description
PDAP1 Polyclonal antibody specifically detects PDAP1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
| PDAP1 | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation | |
| HASPP28, PAP1PDGFA-associated protein 1, PAP28 kDa heat- and acid-stable phosphoprotein, PDGFA associated protein 1, PDGF-associated protein | |
| Produced in rabbits immunized with E. coli-derived Human PDAP1 fragment | |
| 0.1 mL | |
| Signal Transduction | |
| 11333 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunocytochemistry, Immunoprecipitation | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction