Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563903
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563903 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563903 Supplier Invitrogen™ Supplier No. PA563903
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KERYMKMHLA GKTEQAKADL ARLAIIRKQR EEAARKKEEE RKAKDDATLS GKRMQSLSLN K Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

PDGFA-associated protein 1 (PDAP1) is a phosphoprotein that may upregulate the PDGFA-stimulated growth of fibroblasts and also downregulate the mitogenicity of PDGFB. PDGFA in rodents has been shown to bind PDGFA with a low affinity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDAP1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PDAP1
Gene Accession No. Q13442
Gene Alias 28 kDa heat- and acid-stable phosphoprotein; HASPP28; heat- and acid-stable phosphoprotein of 28 kDa; kinase substrate HASPP28; PAP; PAP1; Pdap1; PDGFA associated protein 1; PDGFA-associated protein 1; PDGF-associated protein
Gene Symbols PDAP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PDAP1. Recombinant protein control fragment (Product #RP-103383).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11333
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.