Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PDAP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562166
Description
Immunogen sequence: EKKSLDSDES EDEEDDYQQK RKGVEGLIDI ENPNRVAQTT KKVTQLDLDG PKELSRRERE Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
PDGFA-associated protein 1 (PDAP1) is a phosphoprotein that may upregulate the PDGFA-stimulated growth of fibroblasts and also downregulate the mitogenicity of PDGFB. PDGFA in rodents has been shown to bind PDGFA with a low affinity.
Specifications
PDAP1 | |
Polyclonal | |
Unconjugated | |
PDAP1 | |
28 kDa heat- and acid-stable phosphoprotein; HASPP28; heat- and acid-stable phosphoprotein of 28 kDa; kinase substrate HASPP28; PAP; PAP1; Pdap1; PDGFA associated protein 1; PDGFA-associated protein 1; PDGF-associated protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11333 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q13442 | |
PDAP1 | |
Recombinant protein corresponding to Human PDAP1. Recombinant protein control fragment (Product #RP-103068). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction