Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDAT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144454
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144454 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144454 Supplier Invitrogen™ Supplier No. PA5144454
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PDAT1 Polyclonal Antibody for Western Blot, ELISA

Triacylglycerol formation by an acyl-CoA independent pathway. The enzyme preferentially transfers acyl groups from the sn-2 position of a phospholipid to diacylglycerol, thus forming an sn-1-lysophospholipid. Involved in epoxy and hydroxy fatty acid accumulation in seeds. Has complementary functions with DAG1 that are essential for triacylglycerol synthesis and normal development of both seeds and pollen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDAT1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene PDAT1
Gene Accession No. Q9FNA9
Gene Alias PDAT
Gene Symbols PDAT1
Host Species Rabbit
Immunogen Recombinant Arabidopsis thaliana PDAT1 protein.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 831208
Target Species Plant
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.