Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PDC Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566472
Description
Immunogen sequence: DWRKFKLESQD SDSIPPSKKE ILRQMSSPQS RNGKDSKERV SRKMSIQEYE LI Highest antigen sequence identity to the following orthologs - mouse 89%, rat 91%.
PDC encodes a phosphoprotein, which is located in the outer and inner segments of the rod cells in the retina. This protein may participate in the regulation of visual phototransduction or in the integration of photoreceptor metabolism. It modulates the phototransduction cascade by interacting with the beta and gamma subunits of the retinal G-protein transducin. PDC is a potential candidate gene for retinitis pigmentosa and Usher syndrome type II. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
PDC | |
Polyclonal | |
Unconjugated | |
PDC | |
33 kDa phototransducing protein; 33DPTP; G beta gamma binding protein; MEKA; MEKA protein; Pdc; PHD; PhLOP; PhLP; Phosducin; phosducin-like orphan protein; Phototransducing protein, 33 kDa (phosducin); Protein MEKA; Rod photoreceptor 1; Rpr1; RPR-1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5132 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P20941 | |
PDC | |
Recombinant Human PDC. Recombinant protein control fragment (Product #RP-107146). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction