Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDC-TREM Rat anti-Mouse, BUV615, Clone: 4A6, BD OptiBuild™

Catalog No. BDB751719 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 x 50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751719 1 x 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751719 Supplier BD Biosciences Supplier No. 751719
Add to Cart
Add to Cart

Rat Monoclonal Antibody

PDC-TREM Monoclonal antibody specifically detects PDC-TREM in Mouse samples. It is validated for Flow Cytometry.

TRUSTED_SUSTAINABILITY

Specifications

Antigen PDC-TREM
Applications Flow Cytometry
Classification Monoclonal
Clone 4A6
Concentration 0.2 mg/mL
Conjugate Brilliant Ultraviolet 615
Description Ms PDC-TREM BUV615 4A6 50 μg
Host Species Rat
Immunogen Mouse PDC-TREM Recombinant Protein
Quantity 1 x 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity PDC-TREM
Target Species Mouse
Content And Storage Refrigerate (2 to 8°C)
Isotype IgG2a κ
Reactivity Mouse
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.