Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDC-TREM Rat anti-Mouse, BUV737, Clone: 4A6, BD OptiBuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 751715
Description
PDC-TREM Monoclonal antibody specifically detects PDC-TREM in Mouse samples. It is validated for Flow Cytometry.

Specifications
PDC-TREM | |
Monoclonal | |
0.2 mg/mL | |
Ms PDC-TREM BUV737 4A6 50 μg | |
Mouse PDC-TREM Recombinant Protein | |
RUO | |
PDC-TREM | |
Refrigerate (2 to 8°C) | |
Mouse |
Flow Cytometry | |
4A6 | |
Brilliant Ultraviolet 737 | |
Rat | |
1 x 50 μg | |
Primary | |
Mouse | |
IgG2a κ |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction