Learn More
Description
Specifications
Specifications
| Antigen | PDC-TREM |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 4A6 |
| Concentration | 0.2 mg/mL |
| Conjugate | RB780 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | A530064D06Rik; pdctrem; plasmacytoid dendritic cell-specific receptor; Trem4; triggering receptor expressed on myeloid cells 4 |
| Host Species | Rat |
| Immunogen | Mouse PDC-TREM Recombinant Protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.