Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDC Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA571702
Description
May participate in the regulation of visual phototransduction or in the integration of photoreceptor metabolism. Inhibits the transcriptional activation activity of the cone-rod homeobox CRX.
Specifications
PDC | |
Polyclonal | |
Unconjugated | |
PDC | |
MEKA; PHD; PhLOP; PhLP; phosducin; 33 kDa phototransducing protein; G beta gamma binding protein; phosducin-like orphan protein | |
Rabbit | |
Protein A | |
RUO | |
5132 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.35 mg/mL | |
PBS with 0.09% sodium azide | |
P20941 | |
PDC | |
KLH conjugated synthetic peptide between 59-88 amino acids from the Central region of human PDC | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction