Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDCD1, Mouse anti-Human, Clone: J116, Abnova™

Catalog No. 89999224 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-999-224 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-999-224 Supplier Abnova Corporation Supplier No. MAB16293
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifications

Antigen PDCD1
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone J116
Conjugate Unconjugated
Dilution Flow Cytometry Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (20 ug/mL) Immunohistochemistry (Frozen sections) Immunoprecipitation Western Blot The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.09% sodium azide).
Gene programmed cell death 1
Gene Alias CD279/PD1/SLEB2/hPD-1/hPD-l
Gene Symbols PDCD1
Host Species Mouse
Immunogen Human PDCD1.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5133
Target Species Human
Content And Storage Store at 4°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.