Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDCD1 Mouse anti-Human, Mouse, Clone: OTI26F7, lyophilized, TrueMAB™
SDP

Catalog No. 501668532
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-8532 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-8532 Supplier OriGene Supplier No. CF807864
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

PD-1 (Programmed Cell Death Protein 1, PCDP-1, CD279) is a cell surface receptor approximately 273-290 amino acids in length that inhibits cell-mediated immune responses initiated by T lymphocytes. PD-1 can bind two different but related ligands, PDL-1 and PDL-2. Upon binding to either of these ligands, signals generated by PD-1 inhibit the activation of the immune response in the absence of "danger signals" such as lipopolysaccharide (LPS) associated with bacteria or pathogens. Evidence for this is seen in PD1-null mice who exhibit hyper-activated immune systems and autoimmune diseases. Further, expression of this gene in mice is induced in the thymus when anti-CD3 antibodies are injected and large numbers of thymocytes undergo apoptosis. Mice deficient for this gene bred on a BALB/c background developed dilated cardiomyopathy and died from congestive heart failure.

Specifications

Antigen PDCD1
Applications Flow Cytometry
Classification Monoclonal
Clone OTI26F7
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene PDCD1
Gene Accession No. Q02242, Q15116
Gene Alias CD279, PD-1, PD1, SLEB2, hPD-1, hPD-l, hSLE1
Gene Symbols Pdcd1
Host Species Mouse
Immunogen Full length human recombit protein of human PDCD1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18566, 5133
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.