Learn More
PDCD1 Mouse anti-Human, Mouse, Clone: OTI28F12, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF807865

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
PD-1 (Programmed Cell Death Protein 1, PCDP-1, CD279) is a cell surface receptor approximately 273-290 amino acids in length that inhibits cell-mediated immune responses initiated by T lymphocytes. PD-1 can bind two different but related ligands, PDL-1 and PDL-2. Upon binding to either of these ligands, signals generated by PD-1 inhibit the activation of the immune response in the absence of "danger signals" such as lipopolysaccharide (LPS) associated with bacteria or pathogens. Evidence for this is seen in PD1-null mice who exhibit hyper-activated immune systems and autoimmune diseases. Further, expression of this gene in mice is induced in the thymus when anti-CD3 antibodies are injected and large numbers of thymocytes undergo apoptosis. Mice deficient for this gene bred on a BALB/c background developed dilated cardiomyopathy and died from congestive heart failure.Specifications
PDCD1 | |
Monoclonal | |
Unconjugated | |
PDCD1 | |
CD279, PD-1, PD1, SLEB2, hPD-1, hPD-l, hSLE1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
18566, 5133 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI28F12 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q02242, Q15116 | |
Pdcd1 | |
Full length human recombit protein of human PDCD1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.