Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDCD10 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297685UV
Description
PDCD10 Polyclonal specifically detects PDCD10 in Human samples. It is validated for Western Blot.Specifications
PDCD10 | |
Polyclonal | |
Western Blot | |
apoptosis-related protein 15, cerebral cavernous malformations 3, MGC1212, MGC24477, programmed cell death 10, programmed cell death protein 10, TF-1 cell apoptosis-related protein 15, TFAR15CCM3Cerebral cavernous malformations 3 protein | |
Produced in rabbits immunized with E. coli-derived Human PDCD10 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
11235 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction