Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDCD10 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$207.00 - $682.00
Specifications
Antigen | PDCD10 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 1-5 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:10-1:500 |
Applications | Immunohistochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018647
![]() |
Novus Biologicals
NBP1471950.02MG |
0.02 mg |
Each for $207.00
|
|
|||||
NBP14719501
![]() |
Novus Biologicals
NBP1471950.1MG |
0.1 mg |
Each for $499.50
|
|
|||||
NBP14719502
![]() |
Novus Biologicals
NBP1471950.2MG |
0.2 mg |
Each for $682.00
|
|
|||||
Description
PDCD10 Polyclonal specifically detects PDCD10 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
PDCD10 | |
Western Blot 1-5 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:10-1:500 | |
Polyclonal | |
Purified | |
RUO | |
PBS (pH 7.4) with No Preservative | |
11235 | |
Recombinant human CCM3 | |
Primary | |
Reconstitute in sterile water to a concentration of 0.1-1.0mg/mL. | |
The polyclonal antibody will detect recombinant human CCM-3 in WB and native CCM-3 in IHC |
LYOPH | |
Immunohistochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human | |
apoptosis-related protein 15, cerebral cavernous malformations 3, MGC1212, MGC24477, programmed cell death 10, programmed cell death protein 10, TF-1 cell apoptosis-related protein 15, TFAR15CCM3Cerebral cavernous malformations 3 protein | |
PDCD10 | |
IgG | |
Protein A purified | |
Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title