Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDCD10 Polyclonal Antibody, DyLight™ 488
GREENER_CHOICE

Catalog No. PIPA579792
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579792 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579792 Supplier Invitrogen™ Supplier No. PA579792
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control - Flow: K562 cell.

This gene encodes an evolutionarily conserved protein associated with cell apoptosis. The protein interacts with the serine/threonine protein kinase MST4 to modulate the extracellular signal-regulated kinase (ERK) pathway. It also interacts with and is phosphoryated by serine/threonine kinase 25, and is thought to function in a signaling pathway essential for vascular developent. Mutations in this gene are one cause of cerebral cavernous malformations, which are vascular malformations that cause seizures and cerebral hemorrhages. Multiple alternatively spliced variants, encoding the same protein, have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDCD10
Applications Flow Cytometry
Classification Polyclonal
Concentration 200 μg/mL
Conjugate DyLight 488
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene Pdcd10
Gene Accession No. Q9BUL8
Gene Alias 2410003B13Rik; apoptosis-related protein 15; CCM3; Cerebral cavernous malformations 3 protein; MGC1212; MGC24477; PDCD 10; PDCD10; programmed cell death 10; programmed cell death protein 10; TF-1 cell apoptosis-related protein 15; Tfa15; TFAR15
Gene Symbols Pdcd10
Host Species Rabbit
Immunogen E. coli-derived human PDCD10 recombinant protein (Position: M1-A212).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11235
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.