Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDCD11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553671
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553671 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553671 Supplier Invitrogen™ Supplier No. PA553671
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LTGPHKLEEG EVAMGRVVKV TPNEGLTVSF PFGKIGTVSI FHMSDSYSET PLEDFVPQKV VRCYILSTAD NVLTLSLRSS RTNPETKSKV EDPEINSIQD IKEGQLLRGY VGSIQPHG Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 76%.

PDCD11 is a NF-kappa-B (NFKB1)-binding protein that colocalizes with U3 RNA in the nucleolus and is required for rRNA maturation and generation of 18S rRNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDCD11
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PDCD11
Gene Accession No. Q14690
Gene Alias 1110021I22Rik; ALG4; ALG-4; apoptosis-linked gene 4; apoptosis-linked gene 4 protein; KIAA0185; mKIAA0185; NFBP; NF-kappa-B-binding protein; Pdcd11; Pdcd7; programmed cell death 11; programmed cell death protein 11; programmed cell death protein 7; protein RRP5 homolog; RRP5
Gene Symbols PDCD11
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PDCD11. Recombinant protein control fragment (Product #RP-92052).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22984
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.