Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDCD2 Monoclonal Antibody (DHIC2-5H10)
GREENER_CHOICE

Catalog No. PIMA516135
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516135 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516135 Supplier Invitrogen™ Supplier No. MA516135
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PDCD2 Monoclonal Antibody for IHC (F), ICC/IF, Flow

Essential component of an SCF-type E3 ligase complex, SCF(NIPA), a complex that controls mitotic entry by mediating ubiquitination and subsequent degradation of cyclin B1 (CCNB1). Its cell-cycle-dependent phosphorylation regulates the assembly of the SCF(NIPA) complex, restricting CCNB1 ubiquitination activity to interphase. Its inactivation results in nuclear accumulation of CCNB1 in interphase and premature mitotic entry. May have an antiapoptotic role in NPM-ALK-mediated signaling events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDCD2
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunocytochemistry
Classification Monoclonal
Clone DHIC2-5H10
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation Tissue culture supernatant with 0.05% sodium azide
Gene Pdcd2
Gene Accession No. Q16342
Gene Alias AI528543; hypothetical protein MGC63685; Pcd2; PDCD2; programmed cell death 2; programmed cell death protein 2; RP8; RP-8; zgc:63685; zinc finger MYND domain-containing protein 7; Zinc finger protein Rp-8; ZMYND7
Gene Symbols Pdcd2
Host Species Mouse
Immunogen Human pancreatic enriched duct cells.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5134
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.