Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PDCD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103471
Description
Antibody detects endogenous levels of total PDCD2.
Essential component of an SCF-type E3 ligase complex, SCF(NIPA), a complex that controls mitotic entry by mediating ubiquitination and subsequent degradation of cyclin B1 (CCNB1). Its cell-cycle-dependent phosphorylation regulates the assembly of the SCF(NIPA) complex, restricting CCNB1 ubiquitination activity to interphase. Its inactivation results in nuclear accumulation of CCNB1 in interphase and premature mitotic entry. May have an antiapoptotic role in NPM-ALK-mediated signaling events.
Specifications
PDCD2 | |
Polyclonal | |
Unconjugated | |
Pdcd2 | |
AI528543; hypothetical protein MGC63685; Pcd2; PDCD2; programmed cell death 2; programmed cell death protein 2; RP8; RP-8; zgc:63685; zinc finger MYND domain-containing protein 7; Zinc finger protein Rp-8; ZMYND7 | |
Rabbit | |
Affinity chromatography | |
RUO | |
18567, 5134, 58934 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P46718, P47816, Q16342 | |
Pdcd2 | |
A synthesized peptide derived from human PDCD2(Accession Q16342), corresponding to amino acid residues A6-R56. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction