Learn More
PDCD2L Mouse anti-Human, Clone: OTI4E3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF812472

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Over expression of PDCD2L attenutates TNF-alpha release in Daudi cells. PDCD2L over-expression restrained proliferation of HEK293T cells. DNA/flow cytometry analysis showed that the over-expression of PDCD2L severely delays cell cycle progression at S phase. These all suggest that PDCD2L might play a role in apoptosis.Specifications
PDCD2L | |
Monoclonal | |
Unconjugated | |
PDCD2L | |
programmed cell death 2 like | |
Mouse | |
Affinity Chromatography | |
RUO | |
84306 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI4E3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9BRP1 | |
PDCD2L | |
Full length human recombit protein of human PDCD2L produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.