Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDCD4 Antibody (4C2), DyLight 405 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP318224V
Description
PDCD4 Monoclonal antibody specifically detects PDCD4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
PDCD4 | |
Monoclonal | |
DyLight 405 | |
H731nuclear antigen H731, MGC33046, MGC33047, Neoplastic transformation inhibitor protein, Nuclear antigen H731-like, programmed cell death 4 (neoplastic transformation inhibitor), programmed cell death protein 4, Protein 197/15a | |
Recombinant human PDCD4 (1-469aa) purified from E. coli | |
0.1 mL | |
Apoptosis, Phospho Specific | |
27250 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
4C2 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction