Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDCD4 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298857MFV610
Description
PDCD4 Polyclonal antibody specifically detects PDCD4 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
PDCD4 | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence | |
H731nuclear antigen H731, MGC33046, MGC33047, Neoplastic transformation inhibitor protein, Nuclear antigen H731-like, programmed cell death 4 (neoplastic transformation inhibitor), programmed cell death protein 4, Protein 197/15a | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PDCD4. | |
0.1 mL | |
Apoptosis, Phospho Specific | |
27250 | |
Store at 4C in the dark. | |
IgG |
Immunocytochemistry | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction