Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDCD4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 125871AP150UL

Description
This gene encodes a protein localized to the nucleus in proliferating cells. Expression of this gene is modulated by cytokines in natural killer and T cells. The gene product is thought to play a role in apoptosis but the specific role has not yet been determined. Two transcripts encoding different isoforms have been identified.Specifications
PDCD4 | |
Polyclonal | |
Unconjugated | |
PDCD4 | |
H731, Nuclear antigen H731 like, PDCD4, Protein 197/15a | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18569, 27250, 64031 | |
-20°C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.6 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q53EL6, Q61823, Q9JID1 | |
PDCD4 | |
PDCD4 Fusion Protein Ag3288 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction