Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDCD5 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176988MFV500
Description
PDCD5 Polyclonal antibody specifically detects PDCD5 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
PDCD5 | |
Polyclonal | |
Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
FLJ42784, MGC9294, programmed cell death 5, programmed cell death protein 5, Protein TFAR19, TF-1 cell apoptosis-related protein 19, TFAR19 novel apoptosis-related, TFAR19TF1 cell apoptosis-related gene 19 | |
Antibody was raised against a 21 amino acid synthetic peptide from near the carboxy terminus of human PDCD5. The immunogen is located within the last 50 amino acids of PDCD5. Amino Acid Squence: KFNRRKVMDSDEDDDY | |
0.1 mL | |
Apoptosis | |
9141 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunocytochemistry | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction