Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDCD6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP219741
Description
PDCD6 Polyclonal specifically detects PDCD6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
PDCD6 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
ALG2, ALG-2, Apoptosis-linked gene 2 protein, apoptosis-linked gene-2, FLJ46208, MGC111017, MGC119050, MGC9123, PEF1B, Probable calcium-binding protein ALG-2, programmed cell death 6, programmed cell death protein 6 | |
Rabbit | |
22 kDa | |
0.1 mL | |
Apoptosis | |
10016 | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.62mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
O75340 | |
PDCD6 | |
Recombinant protein encompassing a sequence within the center region of human PDCD6. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Species reactivity based on sequence identity: Xenopus laevis (86%), Zebrafish (88%), Xenopus tropicalis (87%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction