Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDCD7 Antibody (46C713.2.1), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication
$206.50 - $499.50
Specifications
Antigen | PDCD7 |
---|---|
Clone | 46C713.2.1 |
Dilution | Western Blot 1-2 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 28134623) |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056540T
![]() |
Novus Biologicals
NB10056540SS |
0.025 mg |
Each for $206.50
|
|
|||||
NB10056540
![]() |
Novus Biologicals
NB10056540 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
PDCD7 Monoclonal specifically detects PDCD7 in Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
PDCD7 | |
Western Blot 1-2 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 28134623) | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
apoptosis-related protein ES18, ES18programmed cell death protein 7, FLJ54213, hES18, MGC22015, programmed cell death 7, U11/U12 snRNP 59K | |
PDCD7 | |
IgG1 κ | |
Protein G purified |
46C713.2.1 | |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Apoptosis | |
Q8N8D1 | |
10081 | |
The antibody was raised against a synthetic peptide of mouse PDCD-7 protein. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title