Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDCD7 Antibody (46C713.2.1), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056540B
Description
PDCD7 Monoclonal specifically detects PDCD7 in Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
PDCD7 | |
Monoclonal | |
Biotin | |
Q8N8D1 | |
PDCD7 | |
The antibody was raised against a synthetic peptide of mouse PDCD-7 protein. | |
0.1 mL | |
Apoptosis | |
10081 | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, Immunocytochemistry, Immunofluorescence | |
46C713.2.1 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
apoptosis-related protein ES18, ES18programmed cell death protein 7, FLJ54213, hES18, MGC22015, programmed cell death 7, U11/U12 snRNP 59K | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction