Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDCD7 Antibody (SR1994), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322467100UL
Description
PDCD7 Monoclonal antibody specifically detects PDCD7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
PDCD7 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence | |
SR1994 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
apoptosis-related protein ES18, ES18programmed cell death protein 7, FLJ54213, hES18, MGC22015, programmed cell death 7, U11/U12 snRNP 59K | |
A synthesized peptide derived from human PDCD7 (Uniprot #: Q8N8D1) | |
100 μg | |
Apoptosis | |
10081 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction