Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDCD7 Recombinant Rabbit Monoclonal Antibody (8A1Q3)
GREENER_CHOICE

Catalog No. PIMA542671
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542671 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542671 Supplier Invitrogen™ Supplier No. MA542671
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: TE-1, SKOV3, Mouse testis, Mouse thymus, Rat brain. The target is usually found in the following locations: Nucleus. Immunogen sequence: MALPPFFGQG RPGPPPPQPP PPAPFGCPPP PLPSPAFPPP LPQRPGPFPG ASAPFLQPPL ALQPRASAEA SRGGGGAGAF YPVPPPPLPP PPPQCRPFPG.

PDCD7 is a protein with sequence similarity to a mouse protein originally identified in embryonic stem cells. In mouse T-cell lines, this protein appears to be related to glucocorticoid- and staurine-induced apoptotic pathways, and to be linked to ceramide-mediated signaling. These observations suggest that this gene product is involved in specific apoptotic processes in T-cells. This gene encodes a protein with sequence similarity to a mouse protein originally identified in embryonic stem cells. In mouse T-cell lines, this protein appears to be related to glucocorticoid- and staurine-induced apoptotic pathways, and to be linked to ceramide-mediated signaling. These observations suggest that this gene product is involved in specific apoptotic processes in T-cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDCD7
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 8A1Q3
Concentration 0.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Pdcd7
Gene Accession No. Q8N8D1, Q9WTY1
Gene Alias apoptosis-related protein ES18; C80112; ES18; H MGC22015; hES18; LOW QUALITY PROTEIN: programmed cell death protein 7; MGC22015; PDCD7; programmed cell death 7; programmed cell death protein 7; Programmed cell death protein 7-like protein; U11/U12 snRNP 59K
Gene Symbols Pdcd7
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PDCD7 (Q8N8D1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10081, 363082, 50996
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.