Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDCL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561695
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561695 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561695 Supplier Invitrogen™ Supplier No. PA561695
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PPKEESKDEI EEMVLRLQKE AMVKPFEKMT LAQLKEAEDE FDEEDMQAVE TYRKKRLQEW KAL Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.

Phosducin-like proteins (PhLPs) are a conserved family of proteins with thioredoxin-like domains that were initially identified as modulators of G protein signaling. PDCL2 is highly homologous to PDCL and shares an N-terminal helix domain and a C-terminal thioredoxin-fold (Trx-fold) domain. PDCL2 has been shown to interact with the chaperonin CCT and is essential for cytoskeletal protein function and cell cycle progression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDCL2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PDCL2
Gene Accession No. Q8N4E4
Gene Alias 1700010B22Rik; 1700016K07Rik; GCPHLP; MgcPhLP; Pdcl2; phosducin like 2; phosducin-like 2; phosducin-like protein 2; Phosducin-like protein similar 1; protein B; RGD1559712; Tphlp
Gene Symbols PDCL2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PDCL2. Recombinant protein control fragment (Product #RP-100301).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 132954
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.