Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PDCL2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113181
Description
PDCL2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Phosducin-like proteins (PhLPs) are a conserved family of proteins with thioredoxin-like domains that were initially identified as modulators of G protein signaling. PDCL2 is highly homologous to PDCL and shares an N-terminal helix domain and a C-terminal thioredoxin-fold (Trx-fold) domain. PDCL2 has been shown to interact with the chaperonin CCT and is essential for cytoskeletal protein function and cell cycle progression.
Specifications
PDCL2 | |
Polyclonal | |
Unconjugated | |
PDCL2 | |
1700010B22Rik; 1700016K07Rik; GCPHLP; MgcPhLP; Pdcl2; phosducin like 2; phosducin-like 2; phosducin-like protein 2; Phosducin-like protein similar 1; protein B; RGD1559712; Tphlp | |
Rabbit | |
Protein G | |
RUO | |
132954, 498352 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
2 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q8N4E4 | |
PDCL2 | |
Recombinant Human Phosducin-like protein 2 protein (1-241AA). | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction