Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDCL3 Antibody (8F9), Alexa Fluor™ 594, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP242694AF594

Catalog No. NP242694A94


Only null left
Add to Cart

Description

Description

PDCL3 Monoclonal specifically detects PDCL3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

PDCL3
Monoclonal
Alexa Fluor 594
50 mM sodium borate with 0.05% sodium azide
PDCL3
Recombinant human PDCL3 (1-239aa) purified from E. coli
100 μL
Primary
Human
IgG1 κ
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
8F9
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
HTPHLP, IAP-associated factor VIAF1, MGC3062, PHLP3, phosducin-like 3, phosducin-like protein 3, PhPL3, VIAF, VIAF-1, VIAF1PHLP2A, Viral IAP-associated factor 1
Mouse
Protein A purified
Apoptosis
79031.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.