Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDCL3 Antibody (8F9), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242694AF750
Description
PDCL3 Monoclonal specifically detects PDCL3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
PDCL3 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
PDCL3 | |
Recombinant human PDCL3 (1-239aa) purified from E. coli | |
100 μL | |
Primary | |
Human | |
IgG1 κ |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
8F9 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
HTPHLP, IAP-associated factor VIAF1, MGC3062, PHLP3, phosducin-like 3, phosducin-like protein 3, PhPL3, VIAF, VIAF-1, VIAF1PHLP2A, Viral IAP-associated factor 1 | |
Mouse | |
Protein A purified | |
Apoptosis | |
79031.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction