Learn More
PDCL3 Mouse anti-Human, Clone: OTI6G6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809714

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Phosducin-like proteins (PhLPs) are a conserved family of proteins with thioredoxin-like domains that were initially identified as modulators of G protein signaling. PDCL3 is highly homologous to PDCL and shares an N-terminal helix domain and a C-terminal thioredoxin-fold (Trx-fold) domain. Along with the related protein PDCL2, PDCL3 interacts with the chaperonin CCT and modulates CCT-mediated actin and tubulin folding. Modulation of PDCL3 levels by MAPK phosphorylation and RhoA-dependent changes also promote cytoskeletal remodeling.Specifications
PDCL3 | |
Monoclonal | |
Unconjugated | |
PDCL3 | |
HTPHLP, PHLP2A, PHLP3, VIAF, VIAF1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
79031 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI6G6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9H2J4 | |
PDCL3 | |
Full length human recombit protein of human PDCL3 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.