Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDCL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553810
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553810 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553810 Supplier Invitrogen™ Supplier No. PA553810
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IGPLVFGGMN LTRDELEWKL SESGAIMTDL EENPKKPIED VLLSSVRRSV LMKRDSDSEG D Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.

Phosducin-like proteins (PhLPs) are a conserved family of proteins with thioredoxin-like domains that were initially identified as modulators of G protein signaling. PDCL3 is highly homologous to PDCL and shares an N-terminal helix domain and a C-terminal thioredoxin-fold (Trx-fold) domain. Along with the related protein PDCL2, PDCL3 interacts with the chaperonin CCT and modulates CCT-mediated actin and tubulin folding. Modulation of PDCL3 levels by MAPK phosphorylation and RhoA-dependent changes also promote cytoskeletal remodeling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDCL3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PDCL3
Gene Accession No. Q9H2J4
Gene Alias 1110061A19Rik; C80025; HTPHLP; IAP-associated factor VIAF1; PDCL3; PhLP2A; PHLP3; phosducin like 3; phosducin-like 3; phosducin-like protein 3; PhPL3; VIAF; VIAF1; VIAF-1; Viral IAP-associated factor 1
Gene Symbols PDCL3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PDCL3. Recombinant protein control fragment (Product #RP-92035).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79031
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.