Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ PDCL3 Protein

Click to view available options
Quantity:
100μg
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-239 of Human PDCL3 The Recombinant Human PDCL3 Protein is derived from E. coli. The Recombinant Human PDCL3 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
Concentration | 1.0mg/mL |
For Use With (Application) | SDS-PAGE |
Formulation | Liquid. 20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol, 0.1M NaCl |
Molecular Weight (g/mol) | 30.0kDa |
Purification Method | SDS-PAGE |
Quantity | 100μg |
Source | Human |
Storage Requirements | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Regulatory Status | RUO |
Common Name | PDCL3 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction