Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PDD 00017272
GREENER_CHOICE

Catalog No. 70061
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-061 1 mg
70-061-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 70-061 Supplier Tocris Bioscience™ Supplier No. 7006/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent PARG inhibitor

PDD 00017272 is a potent poly (ADP ribose) glycohydrolase (PARG) inhibitor (IC50 values are 4.8 nM in biochemical assay and 9.2 nM in cell based assay).

Specifications

Chemical Name or Material 1-[(2,4-Dimethyl-5-thiazolyl)methyl]-1,2,3,4-tetrahydro-N-(1-methylcyclopropyl)-3-[(2-methyl-5-thiazolyl)methyl]-2,4-dioxo-6-quinazolinesulfonamide
CAS 1945950-20-8
Quantity 1 mg
Target PARG Inhibitors
Molecular Formula C23H25N5O4S3
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.