Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PDDC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA535945
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. Predicted reactivity: Human (93%), Mouse (86%), Rat (86%), Bovine (85%).
The PDDC1 (Parkinson Disease 7 Domain Containing 1) gene encodes a protein that is part of the DJ-1 protein family, known for its role in protecting cells against oxidative stress and neurodegenerative conditions. PDDC1 has been implicated in various cellular processes such as transcription regulation, response to oxidative damage, and cellular homeostasis. It is located on human chromosome 12 and is expressed in several tissues, including the brain. Although its exact function remains to be fully elucidated, PDDC1 shares homology with Parkinson's disease-associated protein DJ-1, suggesting a potential role in neuroprotection. Research indicates that PDDC1 might be involved in neural development and survival, and its dysregulation could contribute to diseases involving oxidative stress and neurodegeneration.
Specifications
| PDDC1 | |
| Polyclonal | |
| Unconjugated | |
| GATD1 | |
| GATD1; Glutamine amidotransferase-like class 1 domain-containing protein 1; Parkinson disease 7 domain containing 1; Parkinson disease 7 domain-containing protein 1; PDDC1; RGD1304836 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 309110, 347862 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8NB37 | |
| GATD1 | |
| Recombinant protein encompassing a sequence within the center region of human PDDC1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction