Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDDC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552613
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552613 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552613 Supplier Invitrogen™ Supplier No. PA552613
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KPICAVGHGV AALCCATNED RSWVFDSYSL TGPSVCELVR APGFARLPLV VEDFVKDSGA CFSASEPDAV HVVLDRHLVT GQNASSTVPA VQNLLFLCGS RK Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 90%.

The PDDC1 (Parkinson Disease 7 Domain Containing 1) gene encodes a protein that is part of the DJ-1 protein family, known for its role in protecting cells against oxidative stress and neurodegenerative conditions. PDDC1 has been implicated in various cellular processes such as transcription regulation, response to oxidative damage, and cellular homeostasis. It is located on human chromosome 12 and is expressed in several tissues, including the brain. Although its exact function remains to be fully elucidated, PDDC1 shares homology with Parkinson's disease-associated protein DJ-1, suggesting a potential role in neuroprotection. Research indicates that PDDC1 might be involved in neural development and survival, and its dysregulation could contribute to diseases involving oxidative stress and neurodegeneration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDDC1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GATD1
Gene Accession No. Q8NB37
Gene Alias GATD1; Glutamine amidotransferase-like class 1 domain-containing protein 1; Parkinson disease 7 domain containing 1; Parkinson disease 7 domain-containing protein 1; PDDC1; RGD1304836
Gene Symbols GATD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PDDC1. Recombinant protein control fragment (Product #RP-89869).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 347862
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.