Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDE10A Antibody (OTI1C9), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273273F
Description
PDE10A Monoclonal specifically detects PDE10A in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
PDE10A | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
PDE10A | |
Full-length protein expressed in 293T cell transfected with human PDE10A expression vector. | |
0.1 mL | |
10846.0 | |
Store at 4°C in the dark. |
Immunohistochemistry | |
OTI1C9 | |
Immunohistochemistry | |
cAMP and cAMP-inhibited cGMP 3′-5′-cyclic phosphodiesterase 10A, dJ416F21.1 (phosphodiesterase 10A), EC 3.1.4, EC 3.1.4.17, EC 3.1.4.35, FLJ11894, FLJ25677, HSPDE10A, phosphodiesterase 10A, phosphodiesterase 10A1 (PDE10A1) | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction